2-Chloro-4-methoxy-3-(trimethylsilyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

18965-1G 18965-500MG 18965-5G 18965-25G
77466-458EA 415.09 USD
77466-458 77479-202 77489-440 77499-796
2-Chloro-4-methoxy-3-(trimethylsilyl)pyridine ≥97%
2-Chloro-4-methoxy-3-(trimethylsilyl)pyridine

Precaución: For research use only

Formula: C₉H₁₄ClNOSi
MW: 215.752 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755731
CAS Number: 777931-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR