1-Chloro-2,4-difluoro-3-(triethylsilyl)benzene ≥97%

Supplier: AOBChem USA

Warning

189358-1G 189358-500MG 189358-5G 189358-25G
77466-454EA 415.09 USD
77466-454 77479-198 77489-436 77499-792
1-Chloro-2,4-difluoro-3-(triethylsilyl)benzene ≥97%
1-Chloro-2,4-difluoro-3-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₂H₁₇ClF₂Si
MW: 262.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30179079
CAS Number: 2182553-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR