1-Bromo-2,4-dichloro-3-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189352-1G 189352-500MG 189352-5G 189352-25G
77466-450EA 415.09 USD
77466-450 77479-194 77489-432 77499-788
1-Bromo-2,4-dichloro-3-(triethylsilyl)benzene ≥95%
1-Bromo-2,4-dichloro-3-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₂H₁₇BrCl₂Si
MW: 340.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30179082
CAS Number: 802984-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR