1-Fluoro-2-methyl-4-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189332-1G 189332-500MG 189332-5G 189332-25G
77466-446EA 415.09 USD
77466-446 77479-190 77489-428 77499-784
1-Fluoro-2-methyl-4-(triethylsilyl)benzene ≥95%
1-Fluoro-2-methyl-4-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₃H₂₁FSi
MW: 224.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860703
CAS Number: 2842852-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR