1-Ethyl-3-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189309-1G 189309-500MG 189309-5G 189309-25G
77466-442EA 415.09 USD
77466-442 77479-186 77489-424 77499-780
1-Ethyl-3-(triethylsilyl)benzene ≥95%
1-Ethyl-3-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₄H₂₄Si
MW: 220.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860683
CAS Number: 18415-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR