1-Ethyl-4-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189299-1G 189299-500MG 189299-5G 189299-25G
77466-438EA 415.09 USD
77466-438 77479-182 77489-420 77499-776
1-Ethyl-4-(triethylsilyl)benzene ≥95%
1-Ethyl-4-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₄H₂₄Si
MW: 220.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860675
CAS Number: 18414-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR