N,N-Dimethyl-3-(triethylsilyl)benzenamine ≥95%

Supplier: AOBChem USA

Warning

189295-1G 189295-500MG 189295-5G 189295-25G
77466-434EA 415.09 USD
77466-434 77479-178 77489-416 77499-772
N,N-Dimethyl-3-(triethylsilyl)benzenamine ≥95%
N,N-Dimethyl-3-(triethylsilyl)benzenamine

Precaución: For research use only

Formula: C₁₄H₂₅NSi
MW: 235.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860672
CAS Number: 1048372-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR