3-(Triethylsilyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

189289-1G 189289-500MG 189289-5G 189289-25G
77466-430EA 415.09 USD
77466-430 77479-174 77489-412 77499-768
3-(Triethylsilyl)-1,1'-biphenyl ≥95%
3-(Triethylsilyl)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₈H₂₄Si
MW: 268.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860666
CAS Number: 51533-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR