5-(Triethylsilyl)-1,3-benzodioxole ≥95%

Supplier: AOBChem USA

Warning

189286-1G 189286-500MG 189286-5G 189286-25G
77466-426EA 415.09 USD
77466-426 77479-170 77489-408 77499-764
5-(Triethylsilyl)-1,3-benzodioxole ≥95%
5-(Triethylsilyl)-1,3-benzodioxole

Precaución: For research use only

Formula: C₁₃H₂₀O₂Si
Storage Temperature: Ambient
MDL Number: MFCD34860664
CAS Number: 1554401-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR