1-Methyl-2-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189277-1G 189277-500MG 189277-5G 189277-25G
77466-418EA 415.09 USD
77466-418 77479-162 77489-400 77499-756
1-Methyl-2-(triethylsilyl)benzene ≥95%
1-Methyl-2-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₃H₂₂Si
MW: 206.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860657
CAS Number: 18412-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR