1-Methoxy-4-(triethylsilyl)benzene ≥97%

Supplier: AOBChem USA

Warning

189266-1G 189266-500MG 189266-5G 189266-25G
77466-414EA 415.09 USD
77466-414 77479-158 77489-396 77499-752
1-Methoxy-4-(triethylsilyl)benzene ≥97%
1-Methoxy-4-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₃H₂₂OSi
MW: 222.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092291
CAS Number: 14311-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR