1,1-Dimethylethyl 3-(2,4-Difluorophenyl)-3-hydroxy-1-azetidinecarboxylate ≥96%

Supplier: AOBChem USA

Warning

189261-1G 189261-500MG 189261-5G 189261-25G
77466-410EA 227.63 USD
77466-410 77479-154 77489-392 77499-748
1,1-Dimethylethyl 3-(2,4-Difluorophenyl)-3-hydroxy-1-azetidinecarboxylate ≥96%
1,1-Dimethylethyl 3-(2,4-Difluorophenyl)-3-hydroxy-1-azetidinecarboxylate

Precaución: For research use only

Formula: C₁₄H₁₇F₂NO₃
MW: 285.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30802683
CAS Number: 1482460-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR