1,1-Dimethylethyl 3-(5-bromo-2-fluorophenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%

Supplier: AOBChem USA

Warning

189243-1G 189243-500MG 189243-5G
77466-406EA 646.84 USD
77466-406 77479-150 77489-388
1,1-Dimethylethyl 3-(5-bromo-2-fluorophenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%
1,1-Dimethylethyl 3-(5-bromo-2-fluorophenyl)-3-hydroxy-1-azetidinecarboxylate

Precaución: For research use only

Formula: C₁₄H₁₇BrFNO₃
MW: 346.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860639
CAS Number: 1515052-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR