1,1-Dimethylethyl 3-(3-cyanophenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%

Supplier: AOBChem USA

Warning

189142-1G 189142-500MG 189142-5G 189142-25G
77466-402EA 415.09 USD
77466-402 77479-146 77489-384 77499-742
1,1-Dimethylethyl 3-(3-cyanophenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%
1,1-Dimethylethyl 3-(3-cyanophenyl)-3-hydroxy-1-azetidinecarboxylate

Precaución: For research use only

Formula: C₁₅H₁₈N₂O₃
MW: 274.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860570
CAS Number: 2387355-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR