1,1-Dimethylethyl 3-(4-chlorophenyl)-3-hydroxy-1-azetidinecarboxylate ≥96%

Supplier: AOBChem USA

Warning

189132-1G 189132-500MG 189132-5G 189132-25G
77466-398EA 381.1 USD
77466-398 77479-142 77489-380 77499-738
1,1-Dimethylethyl 3-(4-chlorophenyl)-3-hydroxy-1-azetidinecarboxylate ≥96%
1,1-Dimethylethyl 3-(4-chlorophenyl)-3-hydroxy-1-azetidinecarboxylate

Precaución: For research use only

Formula: C₁₄H₁₈ClNO₃
MW: 283.75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21596783
CAS Number: 1447464-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR