2-Bromo-1,3-dichloro-4-ethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

189082-1G 189082-500MG 189082-5G 189082-25G
77466-394EA 415.09 USD
77466-394 77479-138 77489-376 77499-734
2-Bromo-1,3-dichloro-4-ethoxybenzene ≥95%
2-Bromo-1,3-dichloro-4-ethoxybenzene

Precaución: For research use only

Formula: C₈H₇BrCl₂O
MW: 269.9505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860545
CAS Number: 2167936-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR