4-Bromo-2-chloropyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

18851-1G 18851-500MG 18851-5G 18851-25G
77466-390EA 166.86 USD
77466-390 77479-134 77489-370 77499-728
4-Bromo-2-chloropyridine-3-boronic acid ≥97%
4-Bromo-2-chloropyridine-3-boronic acid

Precaución: For research use only

Formula: C₅H₄BBrClNO₂
MW: 236.259 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23713091
CAS Number: 2121511-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR