2-(6-Bromo-3-chloro-2-fluorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

18825-1G 18825-500MG 18825-5G 18825-25G
77466-386EA 226.6 USD
77466-386 77479-130 77489-366 77499-722
2-(6-Bromo-3-chloro-2-fluorophenyl)-1,3-dioxolane ≥95%
2-(6-Bromo-3-chloro-2-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇BrClFO₂
MW: 281.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976976
CAS Number: 2432848-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR