3-Chloro-5-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

18778-1G 18778-500MG 18778-5G 18778-25G
77466-382EA 352.26 USD
77466-382 77479-126 77489-362 77499-718
3-Chloro-5-fluoro-4-iodopyridine ≥97%
3-Chloro-5-fluoro-4-iodopyridine

Precaución: For research use only

Formula: C₅H₂ClFIN
Storage Temperature: Ambient
MDL Number: MFCD16610367
CAS Number: 514798-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR