tert-Butyl 3-(3-bromophenyl)-3-hydroxyazetidine-1-carboxylate ≥96%

Supplier: AOBChem USA

Warning

187754-1G 187754-500MG 187754-5G 187754-25G
77466-378EA 410.97 USD
77466-378 77479-122 77489-358 77499-714
tert-Butyl 3-(3-bromophenyl)-3-hydroxyazetidine-1-carboxylate ≥96%
tert-Butyl 3-(3-bromophenyl)-3-hydroxyazetidine-1-carboxylate

Precaución: For research use only

Formula: C₁₄H₁₈BrNO₃
MW: 328.2016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34859583
CAS Number: 1451998-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR