(2-Bromo-6-fluorophenyl)triethylsilane ≥96%

Supplier: AOBChem USA

Warning

187739-1G 187739-500MG 187739-5G 187739-25G
77466-374EA 415.09 USD
77466-374 77479-118 77489-354 77499-712
(2-Bromo-6-fluorophenyl)triethylsilane ≥96%
(2-Bromo-6-fluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₂H₁₈BrFSi
Storage Temperature: Ambient
MDL Number: MFCD30178761
CAS Number: 872545-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR