(5-Bromo-2-fluorophenyl)triethylsilane ≥96%

Supplier: AOBChem USA

Warning

187736-1G 187736-500MG 187736-5G 187736-25G
77466-370EA 415.09 USD
77466-370 77479-114 77489-350 77499-708
(5-Bromo-2-fluorophenyl)triethylsilane ≥96%
(5-Bromo-2-fluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₂H₁₈BrFSi
MW: 289.2592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30178313
CAS Number: 2168094-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR