Ethyl 2-fluoro-3-methoxy-5-methyl-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

187723-1G 187723-500MG 187723-5G
77466-362EA 666.41 USD
77466-362 77479-106 77489-342
Ethyl 2-fluoro-3-methoxy-5-methyl-α-oxobenzeneacetate ≥95%
Ethyl 2-fluoro-3-methoxy-5-methyl-α-oxobenzeneacetate

Precaución: For research use only

Formula: C₁₂H₁₃FO₄
MW: 240.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34859575
CAS Number: 1891985-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR