4-(2-Fluorophenyl)-1-(phenylmethyl)-4-piperidinol ≥95%

Supplier: AOBChem USA

Warning

187090-1G 187090-500MG 187090-5G 187090-25G
77466-358EA 397.58 USD
77466-358 77479-102 77489-338 77499-700
4-(2-Fluorophenyl)-1-(phenylmethyl)-4-piperidinol ≥95%
4-(2-Fluorophenyl)-1-(phenylmethyl)-4-piperidinol

Precaución: For research use only

Formula: C₁₈H₂₀FNO
MW: 285.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28108534
CAS Number: 188863-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR