α-Cyclohexyl-4-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186989-1G 186989-500MG 186989-5G 186989-25G
77466-354EA 415.09 USD
77466-354 77479-098 77489-334 77499-696
α-Cyclohexyl-4-ethylbenzenemethanol ≥95%
α-Cyclohexyl-4-ethylbenzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD11937136
CAS Number: 1178502-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR