α-Cyclohexyl-2,3-dihydro-1,4-benzodioxin-6-methanol ≥95%

Supplier: AOBChem USA

Warning

186986-1G 186986-500MG 186986-5G 186986-25G
77466-350EA 415.09 USD
77466-350 77479-094 77489-330 77499-692
α-Cyclohexyl-2,3-dihydro-1,4-benzodioxin-6-methanol ≥95%
α-Cyclohexyl-2,3-dihydro-1,4-benzodioxin-6-methanol

Precaución: For research use only

Formula: C₁₅H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD11921251
CAS Number: 1181720-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR