α-Cyclohexyl-2,4-dimethylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

186977-1G 186977-500MG 186977-5G 186977-25G
77466-346EA 415.09 USD
77466-346 77479-090 77489-326 77499-688
α-Cyclohexyl-2,4-dimethylbenzenemethanol ≥97%
α-Cyclohexyl-2,4-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD12787072
CAS Number: 1216041-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR