α-Cyclohexyl-2,6-difluoro-4-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186959-1G 186959-500MG 186959-5G 186959-25G
77466-342EA 415.09 USD
77466-342 77479-086 77489-322 77499-684
α-Cyclohexyl-2,6-difluoro-4-methoxybenzenemethanol ≥95%
α-Cyclohexyl-2,6-difluoro-4-methoxybenzenemethanol

Precaución: For research use only

Formula: C₁₄H₁₈F₂O₂
MW: 256.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16356304
CAS Number: 1284265-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR