2,3-Dichloro-α-cyclohexylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186946-1G 186946-500MG 186946-5G 186946-25G
77466-338EA 415.09 USD
77466-338 77479-082 77489-318 77499-680
2,3-Dichloro-α-cyclohexylbenzenemethanol ≥95%
2,3-Dichloro-α-cyclohexylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₆Cl₂O
MW: 259.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222666
CAS Number: 1343027-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR