α-Cyclohexyl-2,6-difluoro-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186919-1G 186919-500MG 186919-5G 186919-25G
77466-334EA 415.09 USD
77466-334 77479-078 77489-314 77499-676
α-Cyclohexyl-2,6-difluoro-3-methylbenzenemethanol ≥95%
α-Cyclohexyl-2,6-difluoro-3-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₁₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD25807687
CAS Number: 1548111-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR