α-Cyclohexyl-4-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186908-1G 186908-500MG 186908-5G 186908-25G
77466-330EA 415.09 USD
77466-330 77479-074 77489-310 77499-672
α-Cyclohexyl-4-(methylthio)benzenemethanol ≥95%
α-Cyclohexyl-4-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₀OS
MW: 236.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34859268
CAS Number: 1598598-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR