α-Cyclohexyl-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186852-1G 186852-500MG 186852-5G
77466-326EA 435.69 USD
77466-326 77479-070 77489-306
α-Cyclohexyl-3-methylbenzenemethanol ≥95%
α-Cyclohexyl-3-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD12153686
CAS Number: 522654-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR