α-Cyclohexyl-1-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

186833-1G 186833-500MG 186833-5G 186833-25G
77466-322EA 415.09 USD
77466-322 77479-066 77489-302 77499-666
α-Cyclohexyl-1-naphthalenemethanol ≥95%
α-Cyclohexyl-1-naphthalenemethanol

Precaución: For research use only

Formula: C₁₇H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD06654250
CAS Number: 94823-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR