6-Bromo-3-chloro-2,4-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18579-1G 18579-500MG 18579-5G 18579-25G
77466-314EA 100.94 USD
77466-314 77479-058 77489-294 77499-658
6-Bromo-3-chloro-2,4-difluorophenylboronic acid ≥97%
6-Bromo-3-chloro-2,4-difluorophenylboronic acid

Precaución: For research use only

Formula: C₆H₃BBrClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28976967
CAS Number: 2121514-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR