3-(2,2-dibromovinyl)quinoline ≥97%

Supplier: AOBChem USA

Warning

185580-1G 185580-500MG 185580-5G
77466-310EA 646.84 USD
77466-310 77479-054 77489-290
3-(2,2-dibromovinyl)quinoline ≥97%
3-(2,2-dibromovinyl)quinoline

Precaución: For research use only

Formula: C₁₁H₇Br₂N
MW: 312.9879 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25954988
CAS Number: 1057216-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR