2-(2,2-dibromovinyl)-5-methylthiophene ≥95%

Supplier: AOBChem USA

Warning

185568-1G 185568-500MG 185568-5G
77466-306EA 497.49 USD
77466-306 77479-050 77489-286
2-(2,2-dibromovinyl)-5-methylthiophene ≥95%
2-(2,2-dibromovinyl)-5-methylthiophene

Precaución: For research use only

Formula: C₇H₆Br₂S
Storage Temperature: Ambient
MDL Number: MFCD28672605
CAS Number: 81294-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR