4-Octylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

184818-1G 184818-500MG 184818-5G 184818-25G
77466-294EA 87.55 USD
77466-294 77479-038 77489-274 77499-642
4-Octylbenzaldehyde ≥97%
4-Octylbenzaldehyde

Precaución: For research use only

Formula: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD00041872
CAS Number: 49763-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR