tert-Butyl 3-(3-chloro-2-fluorophenyl)-3-hydroxypyrrolidine-1-carboxylate ≥96%

Supplier: AOBChem USA

Warning

184426-1G 184426-500MG 184426-5G 184426-25G
77466-290EA 312.09 USD
77466-290 77479-034 77489-268 77499-632
tert-Butyl 3-(3-chloro-2-fluorophenyl)-3-hydroxypyrrolidine-1-carboxylate ≥96%
tert-Butyl 3-(3-chloro-2-fluorophenyl)-3-hydroxypyrrolidine-1-carboxylate

Precaución: For research use only

Formula: C₁₅H₁₉ClFNO₃
MW: 315.7676 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22173454
CAS Number: 1092573-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR