5-Bromo-2-fluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18440-1G 18440-500MG 18440-5G
77466-286EA 544.87 USD
77466-286 77479-030 77489-264
5-Bromo-2-fluoro-3-methoxybenzaldehyde ≥97%
5-Bromo-2-fluoro-3-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22570363
CAS Number: 1779832-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR