4-Bromo-7-fluorobenzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

184389-1G 184389-500MG
77466-282EA 1708.77 USD
77466-282 77479-026
4-Bromo-7-fluorobenzo[d][1,3]dioxole ≥97%
4-Bromo-7-fluorobenzo[d][1,3]dioxole

Precaución: For research use only

Formula: C₇H₄BrFO₂
MW: 219.0078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34683133
CAS Number: 2749366-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR