B-(4-Chlorobenzo[b]thien-2-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

183952-1G 183952-500MG 183952-5G
77466-274EA 657.14 USD
77466-274 77479-018 77489-252
B-(4-Chlorobenzo[b]thien-2-yl)boronic acid ≥95%
B-(4-Chlorobenzo[b]thien-2-yl)boronic acid

Precaución: For research use only

Formula: C₈H₆BClO₂S
MW: 212.46 g/mol
Storage Temperature: Ambient
CAS Number: 2566704-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR