2,2,3,3,3-Pentafluoro-1-(5-fluoro-2-methoxyphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183621-1G 183621-500MG 183621-5G
77466-270EA 590.19 USD
77466-270 77479-014 77489-248
2,2,3,3,3-Pentafluoro-1-(5-fluoro-2-methoxyphenyl)-1-propanone ≥95%
2,2,3,3,3-Pentafluoro-1-(5-fluoro-2-methoxyphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₀H₆F₆O₂
MW: 272.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15145649
CAS Number: 1352208-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR