1-(3,5-Dimethylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183606-1G 183606-500MG 183606-5G
77466-266EA 590.19 USD
77466-266 77479-010 77489-244
1-(3,5-Dimethylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-(3,5-Dimethylphenyl)-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₁H₉F₅O
MW: 252.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15145674
CAS Number: 1352216-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR