2,2,3,3,3-Pentafluoro-1-(4-methoxy-3,5-dimethylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183586-1G 183586-500MG 183586-5G
77466-258EA 668.47 USD
77466-258 77479-002 77489-236
2,2,3,3,3-Pentafluoro-1-(4-methoxy-3,5-dimethylphenyl)-1-propanone ≥95%
2,2,3,3,3-Pentafluoro-1-(4-methoxy-3,5-dimethylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₁F₅O₂
MW: 282.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15145653
CAS Number: 1443314-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR