2,2,3,3,3-Pentafluoro-1-(2,4,6-trifluorophenyl)-1-propanone ≥97%

Supplier: AOBChem USA

Warning

183572-1G 183572-500MG 183572-5G 183572-25G
77466-246EA 415.09 USD
77466-246 77478-990 77489-224 77499-616
2,2,3,3,3-Pentafluoro-1-(2,4,6-trifluorophenyl)-1-propanone ≥97%
2,2,3,3,3-Pentafluoro-1-(2,4,6-trifluorophenyl)-1-propanone

Precaución: For research use only

Formula: C₉H₂F₈O
Storage Temperature: Ambient
MDL Number: MFCD34857751
CAS Number: 1931584-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR