1-(2,3-Difluoro-4-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183548-1G 183548-500MG 183548-5G 183548-25G
77466-242EA 415.09 USD
77466-242 77478-986 77489-220 77499-612
1-(2,3-Difluoro-4-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-(2,3-Difluoro-4-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₀H₅F₇O
MW: 274.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857741
CAS Number: 1963248-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR