1-(2,6-Difluoro-4-methoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183532-1G 183532-500MG 183532-5G 183532-25G
77466-238EA 415.09 USD
77466-238 77478-982 77489-216 77499-608
1-(2,6-Difluoro-4-methoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-(2,6-Difluoro-4-methoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₀H₅F₇O₂
MW: 290.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857728
CAS Number: 1991854-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR