4-(2,2,3,3,3-Pentafluoro-1-oxopropyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

183521-1G 183521-500MG 183521-5G
77466-234EA 594.31 USD
77466-234 77478-978 77489-212
4-(2,2,3,3,3-Pentafluoro-1-oxopropyl)benzonitrile ≥97%
4-(2,2,3,3,3-Pentafluoro-1-oxopropyl)benzonitrile

Precaución: For research use only

Formula: C₁₀H₄F₅NO
Storage Temperature: Ambient
MDL Number: MFCD28105772
CAS Number: 2173105-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR