2,2,3,3,3-Pentafluoro-1-(4-propylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183495-1G 183495-500MG 183495-5G
77466-226EA 668.47 USD
77466-226 77478-970 77489-204
2,2,3,3,3-Pentafluoro-1-(4-propylphenyl)-1-propanone ≥95%
2,2,3,3,3-Pentafluoro-1-(4-propylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₁F₅O
MW: 266.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15145672
CAS Number: 1443353-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR