1-[3-(Dimethylamino)phenyl]-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183486-1G 183486-500MG 183486-5G
77466-222EA 594.31 USD
77466-222 77478-966 77489-200
1-[3-(Dimethylamino)phenyl]-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-[3-(Dimethylamino)phenyl]-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₁H₁₀F₅NO
MW: 267.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103814
CAS Number: 2173103-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR